Pamoic acid, 99%, Thermo Scientific Chemicals
Pamoic acid, 99%, Thermo Scientific Chemicals
Pamoic acid, 99%, Thermo Scientific Chemicals
Pamoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pamoic acid, 99%, Thermo Scientific Chemicals
CAS: 130-85-8 | C23H16O6 | 388.38 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15207.1850 g
Catalog number ALFA15207.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS104-53-0
IUPAC Name3-phenylpropanal
Molecular FormulaC9H10O
InChI KeyYGCZTXZTJXYWCO-UHFFFAOYSA-N
SMILESO=CCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5210-1.5300 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
View more
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
Solubility
Soluble in nitrobenzene, pyridine
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Kristof Prinsen.; Junjie Li.; Hubert Vanbilloen.; Peter Vermaelen.; Ellen Devos.; Luc Mortelmans.; Guy Bormans.; Yicheng Ni.; Alfons Verbruggen. Development and evaluation of a 68Ga labeled pamoic acid derivative for in vivo visualization of necrosis using positron emission tomography. Bioorganic & Medicinal Chemistry. 2010, 18, (14), 5274-5281.
  2. Miao Du.; Zhi-Hui Zhang.; Wei Guo.; Xiao-Juan Fu. Multi-Component Hydrogen-Bonding Assembly of a Pharmaceutical Agent Pamoic Acid with Piperazine or 4,4'-Bipyridyl: A Channel Hydrated Salt with Multiple-Helical Motifs vs a Bimolecular Cocrystal. Cryst. Growth Des. 2009, 9, (4), 1655-1657.