trans-2-Methoxycinnamic acid, 98+%, Thermo Scientific Chemicals
trans-2-Methoxycinnamic acid, 98+%, Thermo Scientific Chemicals
trans-2-Methoxycinnamic acid, 98+%, Thermo Scientific Chemicals
trans-2-Methoxycinnamic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-2-Methoxycinnamic acid, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA15173.065 g
Catalog number ALFA15173.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1633-05-2
IUPAC Namestrontium(2+) carbonate
Molecular FormulaCO3Sr
InChI KeyLEDMRZGFZIAGGB-UHFFFAOYSA-L
SMILES[Sr++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.