3,5-Dichlorobenzoic acid, 99%, Thermo Scientific Chemicals
3,5-Dichlorobenzoic acid, 99%, Thermo Scientific Chemicals
3,5-Dichlorobenzoic acid, 99%, Thermo Scientific Chemicals
3,5-Dichlorobenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dichlorobenzoic acid, 99%, Thermo Scientific Chemicals
CAS: 51-36-5 | C7H4Cl2O2 | 191.007 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15168.065 g
Catalog number ALFA15168.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1623-93-4
IUPAC Name[1,1'-biphenyl]-4-sulfonyl chloride
Molecular FormulaC12H9ClO2S
InChI KeyALBQXDHCMLLQMB-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥96.0%
Melting Point (clear melt)111.0-120.0?C
Identification (FTIR)Conforms
Appearance (Color)White to cream