5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals
5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals
CAS: 99-31-0 | C8H7NO4 | 181.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALFA15074.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS115-77-5
IUPAC Name2,2-bis(hydroxymethyl)propane-1,3-diol
Molecular FormulaC5H12O4
InChI KeyWXZMFSXDPGVJKK-UHFFFAOYSA-N
SMILESOCC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals
Melting point257°C to 265°C
5-Aminoisophthalic acid is a useful biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Aminoisophthalic acid is a useful biochemical for proteomics research.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. S Crapatureanu; R Serbanescu; S B Brevitt; R Kluger. Molecular necklaces. Cross-linking hemoglobin with reagents containing covalently attached ligands. Bioconjugate Chemistry, 1999, 10(6), 1058-1067.
  2. En Tang et. al.Two cobalt(II) 5-aminoisophthalate complexes and their stable supramolecular microporous frameworks. Inorganic Chemistry, 2006, 45(16), 6276-6281.