Methyl propionate, 99%, Thermo Scientific Chemicals
Methyl propionate, 99%, Thermo Scientific Chemicals
Methyl propionate, 99%, Thermo Scientific Chemicals
Methyl propionate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl propionate, 99%, Thermo Scientific Chemicals
CAS: 554-12-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14944.AP500 mL
Catalog number ALFA14944.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS75-07-0
IUPAC Nameacetaldehyde
Molecular FormulaC2H4O
InChI KeyIKHGUXGNUITLKF-UHFFFAOYSA-N
SMILESCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 %
Refractive index1.3310 to 1.3330 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
View more
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.
Solubility
Soluble in water (5g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Rose-Marie Moravie.; Jacques Corset. Conformational behaviour and vibrational spectra of methyl propionate. Chemical Physics Letters. 1974, 26, (2), 210-214.
  2. Giancarlo Albanesi.; Pietro Moggi. Methyl methacrylate by gas phase catalytic condensation of formaldehyde with methyl propionate. Applied Catalysis. 1983, 6, (3), 293-306.