Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals
Used as a nutritional supplement and stabilizer in foods.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFA14894.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialVitamin B12
CAS68-19-9
Recommended StorageKeep cold; Store under Nitrogen
RTECS NumberGG3750000
EINECS Number200-680-0
View more
Vitamin B12 is used as a nutritional supplement and stabilizer in foods. It is also used as a medicine and animal feed supplement. It plays an important role in the synthesis of myelin. It is also used as an antipernicious anemia factor. Further, it is used to treat cyanide poisoning and hereditary deficiency of transcobalamin II.