2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-4-methylthiazole, 98+%, Thermo Scientific Chemicals
CAS: 1603-91-4 | C4H6N2S | 114.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number ALFA14847.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight114.17