n-Dodecane, ≥99%, Thermo Scientific Chemicals
n-Dodecane, ≥99%, Thermo Scientific Chemicals
n-Dodecane, ≥99%, Thermo Scientific Chemicals
n-Dodecane, ≥99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Dodecane, ≥99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
25 g
Catalog number ALFA14834.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS931-36-2
IUPAC Name2-ethyl-5-methyl-1H-imidazole
Molecular FormulaC6H10N2
InChI KeyULKLGIFJWFIQFF-UHFFFAOYSA-N
SMILESCCC1=NC=C(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.4990-1.5050 @ 20?C
FormLumps or clear liquid or viscous liquid as melt
View more
n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.
Solubility
Miscible with ethyl alcohol, ethyl ether, acetone, chloroform and carbon tetrachloride. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Pei, Y.; Hawkes, E. R.; Kook, S.; Goldin, G. M.; Lu, T. Modelling n-dodecane spray and combustion with the transported probability density function method. Combust. Flame 2015, 162 (5), 2006-2019.
  2. Vo, Q. N.; Hawkins, C. A.; Dang, L. X.; Nilsson, M.; Nguyen, H. D. Computational Study of Molecular Structure and Self-Association of Tri-n-butyl Phosphates in n-Dodecane. J. Phys. Chem. B 2015, 119 (4), 1588-1597.