Diphenyl phthalate, 98%, Thermo Scientific Chemicals
Diphenyl phthalate, 98%, Thermo Scientific Chemicals
Diphenyl phthalate, 98%, Thermo Scientific Chemicals
Diphenyl phthalate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diphenyl phthalate, 98%, Thermo Scientific Chemicals
CAS: 84-62-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14449.1425 g
Catalog number ALFA14449.14
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS81-54-9
IUPAC Name1,2,4-trihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O5
InChI KeyBBNQQADTFFCFGB-UHFFFAOYSA-N
SMILESOC1=CC(O)=C2C(=O)C3=CC=CC=C3C(=O)C2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=85 %
Appearance (Color)Dark red to orange-brown to brown
Appearance (Form)Powder
UV Lambda max470 to 490 nm (10 mg/l methanol)