3,5-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
3,5-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
3,5-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
3,5-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
CAS: 1132-21-4 | C9H10O4 | 182.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14374.22100 g
Catalog number ALFA14374.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3,5-Dimethoxybenzoic acid
CAS1132-21-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c