Copper(II) nitrate hemi(pentahydrate), 98%
Copper(II) nitrate hemi(pentahydrate), 98%
Copper(II) nitrate hemi(pentahydrate), 98%
Copper(II) nitrate hemi(pentahydrate), 98%
Thermo Scientific Chemicals
Copper(II) nitrate hemi(pentahydrate), 98%
CAS: 19004-19-4 | Cu2H10N4O17 | 465.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14004.36500 g
Catalog number ALFA14004.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS13450-84-5
IUPAC Namegadolinium(3+) hexahydrate trichloride
Molecular FormulaCl3GdH12O6
InChI KeyPNYPSKHTTCTAMD-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
Solubility
Soluble in water, ammonia and ethanol
Notes
Incompatible with reducing agents, organic materials and powdered metals.
RUO – Research Use Only