o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA13602.30250 g
Catalog number ALFA13602.30
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Materialo-Toluidine
CAS95-53-4
Health Hazard 1H227-H301+H331-H312-H315-H318-H341-H350
Health Hazard 2GHS H Statement
H301-H331-H350-H227-H319
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
Combustible liquid.
Causes serious eye irritation.
Health Hazard 3P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P332+P313-P362-P370+P378q-P501c
View more
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.
Solubility
Soluble in water(15g/L).
Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Aulis Hyvarinen.; Esko A. Nikkila. Specific determination of blood glucose with o-toluidine. Clinica Chimica Acta. 1962, 7, (1), 140-143.
  2. AL-Othmana, Z.A.; Mu. Naushad.; Inamuddin. Organic-inorganic type composite cation exchanger poly-o-toluidine Zr(IV) tungstate: Preparation, physicochemical characterization and its analytical application in separation of heavy metals. Chemical Engineering Journal. 2011, 72, (1), 369-375.