Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piperine, 98%, Thermo Scientific Chemicals
CAS: 94-62-2 | C17H19NO3 | 285.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA13510.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7705-08-0
IUPAC Nameiron(3+) trichloride
Molecular FormulaCl3Fe
InChI KeyRBTARNINKXHZNM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.
Solubility
Soluble in water(0.04g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Vladimir Badmaeva.; Muhammed Majeeda.; Lakshmi Prakash. Piperine derived from black pepper increases the plasma levels of coenzyme q10 following oral supplementation ☆. The Journal of Nutritional Biochemistry. 2000, 11, (2), 109-113.
  2. Junhui Liu.; Ying Bi.; Rong Luo.; Xia Wu. Simultaneous UFLC-ESI-MS/MS determination of piperine and piperlonguminine in rat plasma after oral administration of alkaloids from Piper longum L.: Application to pharmacokinetic studies in rats. Journal of Chromatography B. 2011, 879, (27), 2885-2890.