Methyl 4-amino-2-methoxybenzoate, 98%, Thermo Scientific Chemicals
Methyl 4-amino-2-methoxybenzoate, 98%, Thermo Scientific Chemicals
Methyl 4-amino-2-methoxybenzoate, 98%, Thermo Scientific Chemicals
Methyl 4-amino-2-methoxybenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 4-amino-2-methoxybenzoate, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA13239.065 g
Catalog number ALFA13239.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS754-05-2
IUPAC Nameethenyltrimethylsilane
Molecular FormulaC5H12Si
InChI KeyGCSJLQSCSDMKTP-UHFFFAOYSA-N
SMILESC[Si](C)(C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.3895 to 1.3915 (20°C, 589 nm)