Naphthalene, 99+%, Thermo Scientific Chemicals
Naphthalene, 99+%, Thermo Scientific Chemicals
Naphthalene, 99+%, Thermo Scientific Chemicals
Naphthalene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99+%, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13188.0B1000 g
Catalog number ALFA13188.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS54-64-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)≥96.0%
Identification (FTIR)Conforms
Loss on Drying≤0.5%
FormCrystalline powder
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.
Solubility
Soluble in benzene, toluene, chloroform, carbon disulfide, ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Reacts with Na metal in 1,2-dimethoxyethane to give Na naphthalenide. This radical anion is a powerful-electron donor, e.g. in reductive dehalogenation of vic-dihalides: J. Chem. Soc., Chem. Commun., 78 (1969), and in reductive cleavage of tosylates to alcohols: J. Am. Chem. Soc., 88, 1581 (1966).
  2. Li naphthalenide is a useful, non-nucleophilic reagent for the conversion of carboxylic acids to their dianions in cases where LDA could interfere: J. Org. Chem., 45, 1106 (1980). Similarly, has been found preferable to LDA in the lithiation of allylic nitriles: Synthesis, 358 (1981).
  3. Carboxylic acids react with alkyl, alkenyl or aryl chlorides in the presence of Li metal and a catalytic amount of naphthalene in THF to give ketones in fair to good yields: J. Org. Chem., 61, 6058 (1996).
  4. Hwang, Y. J.; Earmme, T.; Courtright, B. A.; Eberle, F. N.; Jenekhe, S. A. n-Type Semiconducting Naphthalene Diimide-Perylene Diimide Copolymers: Controlling Crystallinity, Blend Morphology, and Compatibility Toward High-Performance All-Polymer Solar Cells. J. Am. Chem. Soc. 2015, 137 (13), 4424-4434.
  5. Attah, I. K.; Platt, S. P.; Meot-Ner, M.; El-Shall, M. S.; Peverati, R.; Head-Gordon, M. What Is the Structure of the Naphthalene-Benzene Heterodimer Radical Cation? Binding Energy, Charge Delocalization, and Unexpected Charge-Transfer Interaction in Stacked Dimer and Trimer Radical Cations. J. Phys. Chem. Lett. 2015, 6 (7), 1111-1118.