2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
CAS: 60-56-0 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA13094.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-40+100 mesh (150 - 425 micron)
Trace Metalsless than 0.5% trace metals
Possesses antioxidant properties