2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
CAS: 60-56-0 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA13094.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS63503-60-6
IUPAC Name(3-chlorophenyl)boronic acid
Molecular FormulaC6H6BClO2
InChI KeySDEAGACSNFSZCU-UHFFFAOYSA-N
SMILESOB(O)C1=CC(Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-green
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
HPLC>=96.0 %
Possesses antioxidant properties