2,5-Dimethylfuran, 98+%, Thermo Scientific Chemicals
2,5-Dimethylfuran, 98+%, Thermo Scientific Chemicals
2,5-Dimethylfuran, 98+%, Thermo Scientific Chemicals
2,5-Dimethylfuran, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimethylfuran, 98+%, Thermo Scientific Chemicals
CAS: 625-86-5 | C6H8O | 96.129 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12833.1425 g
Catalog number ALFA12833.14
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2459-09-8
IUPAC Namemethyl pyridine-4-carboxylate
Molecular FormulaC7H7NO2
InChI KeyOLXYLDUSSBULGU-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear orange to brown
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5110 to 1.5140 (20°C, 589 nm)
Appearance (Form)Liquid
2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.
Solubility
Slightly miscible with water. Miscible with ethanol and fats.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. Hu, L.; Lin, L.; Liu, S. Chemoselective Hydrogenation of Biomass-Derived 5-Hydroxymethylfurfural into the Liquid Biofuel 2,5-Dimethylfuran. Ind. Eng. Chem. Res. 2014, 53 (24), 9969-9978.
  2. Kong, X.; Zhu, Y.; Zheng, H.; Dong, F.; Zhu, Y.; Lia, Y-W. Switchable synthesis of 2,5-dimethylfuran and 2,5-dihydroxymethyltetrahydrofuran from 5-hydroxymethylfurfural over Raney Ni catalyst. RSC Adv. 2014, 4 (105), 60467-60472.
  3. Zhang, Q.; Chen, G.; Zheng, Z.; Liu, H.; Xu, J.; Yao, M. Combustion and emissions of 2,5-dimethylfuran addition on a diesel engine with low temperature combustion. Fuel 2013, 103, 730-735.
  4. Wijaya, Y. P.; Suh, D. J.; Jae, J. Production of renewable p-xylene from 2,5-dimethylfuran via Diels-Alder cycloaddition and dehydrative aromatization reactions over silica-alumina aerogel catalysts. Catal. Commun. 2015, 70, 12-16.