2,5-Dimethylaniline, 98+%, Thermo Scientific Chemicals
2,5-Dimethylaniline, 98+%, Thermo Scientific Chemicals
2,5-Dimethylaniline, 98+%, Thermo Scientific Chemicals
2,5-Dimethylaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dimethylaniline, 98+%, Thermo Scientific Chemicals
CAS: 95-78-3 | C8H11N | 121.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12682.22100 g
Catalog number ALFA12682.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2,5-Dimethylaniline
Melting Point15°C
CAS95-78-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
View more
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.
Solubility
Soluble in water <0.1 g/100 mL at 18°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. V Bavastrello.; E Stura.; S Carrara.; V Erokhin. Poly (2, 5-dimethylaniline)-MWNTs nanocomposite: a new material for conductometric acid vapors sensor. Sensors and Actuators B: Chemical. 2004, 98, (2-3), 247-253.
  2. EM Genies.; P Noel. Oxidation of 2, 5-dimethylaniline in NH 4 F· 2.35 HF medium Characteristics of the resulting conducting polymer. Journal of electroanalytical chemistry and interfacial electrochemistry. 1990, 296, (2), 473-490.