1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals
1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals
1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals
1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals
CAS: 86-93-1 | C7H6N4S | 178.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA12656.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥99.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)<250 ppm
Refractive Index1.4225-1.4265 @ 20?C
View more
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.
Solubility
Soluble in 5% ethanol, water (partly), acetone, chloroform, and methanol.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. R Cea-Olivares.; OJ Sandoval.; G Espinosa-Perez. New organotin(IV) derivatives of 1-phenyl-1H-tetrazole-5-thiol. Crystal and molecular structure of 1∞[Bz3SnSCN4Ph], a polymer built through secondary intermolecular Sn⋯N bonds. Polyhedron. 2008, 13, (19), 2809-2818.
  2. C Ma.; J Sun. Hydrogen bonding and π-π stacking interaction in the coordination of sulfur and nitrogen donor heterocycle to organotin(IV): syntheses and crystal structures of di- and triorganotin(IV) derivatives with 1-(4-hydroxyphenyl)-1H-tetrazole-5-thiol. Polyhedron. 2004, 23, (9), 2809-2818.
  3. In combination with t-butyl isocyanide, promotes the lactonization of long-chain hydroxy acids: Angew. Chem. Int. Ed., 20, 771 (1981).