2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
CAS: 149-57-5 | C8H16O2 | 144.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFA12644.AP
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS89343-06-6
IUPAC Nameethynyltris(propan-2-yl)silane
Molecular FormulaC11H22Si
InChI KeyKZGWPHUWNWRTEP-UHFFFAOYSA-N
SMILESCC(C)[Si](C#C)(C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless
FormLiquid
Assay (GC)>96.0%
Refractive Index1.4500-1.4550 @ 20?C
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.
Solubility
Miscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Sun, T.; Qi, M.; Fu, R. Perhydroxylcucurbit[6]uril as a highly selective gas chromatographic stationary phase for analytes of wide-ranging polarity. J. Sep. Sci. 2015, 38 (5), 821-824.
  2. Mochalski, P.; Theurl, M.; Sponring, A.; Unterkofler, K.; Kirchmair, R.; Amann, A. Analysis of Volatile Organic Compounds Liberated and Metabolised by Human Umbilical Vein Endothelial Cells (HUVEC) In Vitro. Cell Biochem. Biophys. 2015, 71 (1), 323-329.