4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
CAS: 92-66-0 | C12H9Br | 233.108 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFA12638.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1192-62-7
IUPAC Name1-(furan-2-yl)ethan-1-one
Molecular FormulaC6H6O2
InChI KeyIEMMBWWQXVXBEU-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
GC>=98.5 %
Appearance (Form)Liquid or low melting solid
Refractive index1.5060 to 1.5080 (20°C, 589 nm)
Infrared spectrumConforms
View more
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.
Solubility
Insoluble in water.
Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. A Parkinson; and S Safe. Cytochrome P-450-mediated metabolism of biphenyl and the 4-halobiphenyls. Biochemical Pharmacology. 1982, 31(10), 1849-1856.
  2. Rusling JF, et al. Electrocatalytic reactions in organized assemblies: Part I. Reduction of 4-bromobiphenyl in cationic and non-ionic micelles. J. Electroanal. Chem. Interfac. Electrochem. 1988, 240(1), 201-216.