Terephthalic acid, 98+%, Thermo Scientific Chemicals
Terephthalic acid, 98+%, Thermo Scientific Chemicals
Terephthalic acid, 98+%, Thermo Scientific Chemicals
Terephthalic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terephthalic acid, 98+%, Thermo Scientific Chemicals
CAS: 100-21-0 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Catalog number ALFA12527.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS6269-89-2
IUPAC Name1-(4-nitrophenyl)piperazine
Molecular FormulaC10H13N3O2
InChI KeyVWOJSRICSKDKAW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point127-134?C
FormPowder
Identification (FTIR)Conforms
Appearance (Color)Yellow to orange to brown
Assay (GC)≥97.5%
Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.
Solubility
slightly soluble in water (0,017 g/L at 25°C)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It should be stored and used in molten form.
RUO – Research Use Only
General References:
  1. Hwa-Jeong Lee. ; Se-Jin Oh. ; Ja-Young Choi .; Jin Won Kim.; Jungwan Han.; Loon-Seng Tan.; Jong-Beom Baek. In Situ Synthesis of Poly(ethylene terephthalate) (PET) in Ethylene Glycol Containing Terephthalic Acid and Functionalized Multiwalled Carbon Nanotubes (MWNTs) as an Approach to MWNT/PET Nanocomposites. Chem. Mater. 2005, 17 (20),5057-5064.
  2. M. Bailey.; C. J. Brown. The crystal structure of terephthalic acid. Acta Cryst. 1967, 22 (3),387-391 .