Blue Tetrazolium chloride, Thermo Scientific Chemicals
Blue Tetrazolium chloride, Thermo Scientific Chemicals
Blue Tetrazolium chloride, Thermo Scientific Chemicals
Blue Tetrazolium chloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Blue Tetrazolium chloride, Thermo Scientific Chemicals
CAS: 1871-22-3 | C40H36Cl2N8O2 | 731.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA12502.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBlue Tetrazolium chloride
CAS1871-22-3
Melting Point∼255°C (decomposition)
Recommended StorageAmbient temperatures
EINECS Number217-488-8
View more
Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.
Solubility
Soluble in hot water (5 mg/ml), methanol (50 mg/ml), ethanol, and chloroform. Insoluble in acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. C.K.Jue; P.N.Lipke. Determination of reducing sugars in the nanomole range with tetrazolium blue chloride. Journal of Biochemical and Biophysical Methods. 1985, 11, (2-3)109-115
  2. Vincent A.DeBari; Mark A.Needle. Cation complexed DNA. A novel class of immunoadsorbents for anti-DNA antibodies. Journal of Immunological Methods. 1981, 40, (1)89-94
  3. Reagent for identification of reducing sugars.
  4. For use in the oxidation of steroids, see: J. Chem. Soc., Chem. Commun., 2687 (1988).