1,5-Dichloropentane, 98%, Thermo Scientific Chemicals
1,5-Dichloropentane, 98%, Thermo Scientific Chemicals
1,5-Dichloropentane, 98%, Thermo Scientific Chemicals
1,5-Dichloropentane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,5-Dichloropentane, 98%, Thermo Scientific Chemicals
CAS: 628-76-2 | C5H10Cl2 | 141.035 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA12433.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.
Solubility
Miscible with ethanol, ether and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Chorazewski, M.; Grzybowski, A.; Paluch, M. Isobaric Thermal Expansion of Compressed 1,4-Dichlorobutane and 1-Bromo-4-chlorobutane: Transitiometric Results and a Novel Application of the General Density Scaling-Based Equation of State. Ind. Eng. Chem. Res. 2015, 54 (24), 6400-6407.
  2. Nicola, F. P. D.; Lanzi, M.; Marchetti, F.; Pampaloni, G.; Zacchini, S. Is bond stretch isomerism in mononuclear transition metal complexes a real issue? The misleading case of the MoCl5/tetrahydropyran reaction system. Dalton Trans. 2015, 44 (28), 12653-12659.