Hematoxylin hydrate, 96% (dry wt.), water ca 6%, Thermo Scientific Chemicals
Hematoxylin hydrate, 96% (dry wt.), water ca 6%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hematoxylin hydrate, 96% (dry wt.), water ca 6%, Thermo Scientific Chemicals
CAS: 517-28-2 | C{1}{6}H{1}{4}O{6}^.xH{2}O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ALFA12431.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Melting Pointca 200°C (decomposition)
CAS517-28-2
Quantity25 g
Molecular FormulaC{1}{6}H{1}{4}O{6}^.xH{2}O
SynonymC.I. 75290;Natural Black 1
View more
Extracted from the bark of the logwood tree
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Extracted from the bark of the logwood tree