Methyl benzoate, 99%, Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
CAS: 93-58-3 | C8H8O2 | 136.15 g/mol
Have Questions?
Catalog NumberdimQuantity
ALFA12404.30
Catalog number ALFA12404.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight136.15
Unit Size
Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Cerreta, M. M.; Furton, K. G. An assessment of detection canine alerts using flowers that release methyl benzoate, the cocaine odorant, and an evaluation of their behavior in terms of the VOCs produced. Forensic Sci. Int. 2015, 251, 107-114.
  2. Maiti, K. S. Vibrational spectroscopy of Methyl benzoate. Phys. Chem. Chem. Phys. 2015, 17 (30), 19735-19744.
  3. Sun, H.; Page, M. I.; Atherton, J. H.; Hall, A. Kinetics of the conversion of methyl benzoate to benzamide by the alumina catalysed reaction with liquid ammonia at 120°C. Catal. Sci. Technol. 2014, 4 (11), 3870-3878.