S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
S-Methyl thioacetate, 98+%, Thermo Scientific Chemicals
CAS: 1534-08-3 | C3H6OS | 90.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12400.1425 g
Catalog number ALFA12400.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialS-Methyl thioacetate
CAS1534-08-3
Health Hazard 1H225-H311+H331-H319
Health Hazard 2GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P363-P370+P378q-P501c