(S)-(-)-Nicotine, 98%, Thermo Scientific Chemicals
(S)-(-)-Nicotine, 98%, Thermo Scientific Chemicals
(S)-(-)-Nicotine, 98%, Thermo Scientific Chemicals
(S)-(-)-Nicotine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(S)-(-)-Nicotine, 98%, Thermo Scientific Chemicals
L-Nicotine, CAS # 54-11-5, has been shown in laboratory studies to alleviate smoking habits. It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine.
Have Questions?
Catalog NumberQuantity
ALFA12398.1425 g
Catalog number ALFA12398.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7647-10-1
IUPAC Namepalladium(2+) dichloride
Molecular FormulaCl2Pd
InChI KeyPIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)Pd 9.0-11.0%
(S)-(-)-Nicotine is utilized as a therapeutic agent to eliminate smoking with the damage it does to health. It is widely used as a drug to enhance the memory or other cognition functions, which is second to caffeine. Lesser levels of nicotine given in the form of gum, may help to eliminate smoking.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• L-Nicotine is a naturally occuring alkaloid in the nightshade family of plants
• It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine
Applications
• L-Nicotine has been shown in studies to alleviate smoking habits
• It has also been shown to enhance cognitive functions such as memory
• Studies show that lower levels of nicotine in the form of gum may also help eliminate smoking habits
RUO – Research Use Only
General References:
  1. Ortega, P. G. R.; Montejo, M.; González, J. J. L. Vibrational Circular Dichroism and Theoretical Study of the Conformational Equilibrium in (-)-S-Nicotine. Angew. Chem. 2015, 16 (2), 342-352.
  2. Zhou, T.; Jørgensen, L.; Mattebjerg, M. A.; Chronakis, I. S.; Ye, L. Molecularly imprinted polymer beads for nicotine recognition prepared by RAFT precipitation polymerization: a step forward towards multi-functionalities. RSC Adv. 2014, 4,30292-30299.