Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals
Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals
Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals
Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals
CAS: 41468-25-1 | C8H10NO6P | 247.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12323.065 g
Catalog number ALFA12323.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS42201-71-8
IUPAC Namemethyl 3-(trimethylsilyl)prop-2-ynoate
Molecular FormulaC7H12O2Si
InChI KeyXXGHAMBKHIDZDC-UHFFFAOYSA-N
SMILESCOC(=O)C#C[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4380-1.4420 @ 20?C
Assay (GC)≥97.5%
FormLiquid
Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.
Solubility
Soluble in alkaline solutions. Sparingly soluble in water.
Notes
Light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
RUO – Research Use Only
General References:
  1. Zhao, X. Y.; Zhu, Y. J.; Lu, B. Q.; Chen, F.; Qi, C.; Zhao, J.; Wu, J. Hydrothermal synthesis of hydroxyapatite nanorods using pyridoxal-5'-phosphate as a phosphorus source. Mater. Res. Bull. 2014, 55, 67-70.
  2. Vilanova, B.; Gallardo, J. M.; Caldés, C.; Adrover, M.; Ortega-Castro, J.; Munoz, F.; Donoso, J. Formation of Schiff bases of O-phosphorylethanolamine and O-phospho-d, l-serine with pyridoxal 5'-phosphate. Experimental and theoretical studies. J. Phys. Chem. A 2012, 166 (8), 1897-1905.