2-Chlorobenzimidazole, 97%, Thermo Scientific Chemicals
2-Chlorobenzimidazole, 97%, Thermo Scientific Chemicals
2-Chlorobenzimidazole, 97%, Thermo Scientific Chemicals
2-Chlorobenzimidazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobenzimidazole, 97%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA12193.065 g
Catalog number ALFA12193.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7681-65-4
IUPAC Nameλ¹-copper(1+) iodide
Molecular FormulaCuI
InChI KeyLSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES[Cu+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric31.0 to 34.0 % (Cu)
Appearance (Color)White to grey to light brown
Appearance (Form)Powder
Total trace metal impurities=<50 ppm
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents, alkali metals, bases.
RUO – Research Use Only
General References:
  1. Ryuichi Iemura; Tsuneo Kawashima; Toshikazu Fukuda; Keizo Ito; Goro Tsukamoto. Synthesis of 2-(4-substituted-1-piperazinyl)benzimidazoles as H1-antihistaminic agents. J. Med. Chem. 1986, 29 (7), 1178-1183.
  2. D. Harrison; J. T. Ralph and A. C. B. Smith. Some 1- and 2-halogenobenzimidazoles. J. Chem. Soc. 1963, 2930-2937.