1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals
1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals
1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals
1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals
CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
100 g
250 g
Catalog number ALFA12160.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS353-43-5
IUPAC Name(methylsulfanyl)methane; trifluoroborane
Molecular FormulaC2H6BF3S
InChI KeyBRWZPVRDOUWXKE-UHFFFAOYSA-N
SMILESCSC.FB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange
FormLiquid
Assay (Aqueous acid-base Titration)94-106%
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
Solubility
Soluble in water 0.045g/L.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Harry A.J. Oonka; Peter R. van der Lindeb; Jan Huinink; Jacobus G. Blok. Representation and assessment of vapor pressure data; a novel approach applied to crystalline 1-bromo-4-chlorobenzene, 1-chloro-4-iodobenzene, and 1-bromo-4-iodobenzene. The Journal of Chemical Thermodynamics. 1998, 30 (7), 897-907.
  2. Vincenzo Calò; Angelo Nacci; Antonio Monopoli Dr.; and Pietro Cotugno. Heck Reactions with Palladium Nanoparticles in Ionic Liquids: Coupling of Aryl Chlorides with Deactivated Olefins. Angewandte Chemie. 2009, 121 (33), 6217-6219.