p-Toluenesulfonamide, 98+%, Thermo Scientific Chemicals
p-Toluenesulfonamide, 98+%, Thermo Scientific Chemicals
p-Toluenesulfonamide, 98+%, Thermo Scientific Chemicals
p-Toluenesulfonamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonamide, 98+%, Thermo Scientific Chemicals
CAS: 70-55-3 | C7H9NO2S | 171.214 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12156.30250 g
Catalog number ALFA12156.30
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS194491-31-1
IUPAC Nametetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12N2Na4O8
InChI KeyUEUXEKPTXMALOB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
Infrared spectrumConforms
Loss on drying5 % to 15 % (160°C)
Titration Complexometric>=97.5 % (on dry substance)
p-Toluene sulfonamide was used to prepare the precursor required for synthesis of ethyl 6-phenyl-1-tosyl-1,2,5,6-tetrahydropyridine-3-carboxylate. It act as a derivative of ammonia activated to alkylation by alkyl halides is exemplified by the synthesis of N-tosyl-2,3-dihydroisoindole from o-xylylene dibromide. It is a precursor of N-tosyl imines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Toluene sulfonamide was used to prepare the precursor required for synthesis of ethyl 6-phenyl-1-tosyl-1,2,5,6-tetrahydropyridine-3-carboxylate. It act as a derivative of ammonia activated to alkylation by alkyl halides is exemplified by the synthesis of N-tosyl-2,3-dihydroisoindole from o-xylylene dibromide. It is a precursor of N-tosyl imines.
Solubility
Soluble in water.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Kui Lu; Ohyun Kwon. Phosphine-Catalyzed [4+2] Annulation: Synthesis of Ethyl 6-Phenyl-1-Tosyl-1,2,5,6-Tetrahydropyridine-3-Carboxylate. Organic Syntheses. 2009, 2009 (86), 212-224.
  2. D. S. Tarbell; Clay Weaver. The Condensation of Sulfoxides with p-Toluenesulfonamide and Substituted Acetamides. J. Am. Chem. Soc. 1941, 63 (11), 2939-2942.
  3. Use of p-toluenesulfonamide as a derivative of ammonia activated to alkylation by alkyl halides is exemplified by the synthesis of N-tosyl-2,3-dihydroisoindole from o-xylylene dibromide: Org. Synth. Coll., 5, 1064 (1973). Similarly, has been used as the precursor of nitrogen-containing crown ethers; see, e.g.: J. Org. Chem., 53, 5292 (1988).
  4. Precursor of N-tosyl imines, which are useful intermediates in synthesis: Synlett, 2097 (2003).