Vanillic acid, 98%, Thermo Scientific Chemicals
Vanillic acid, 98%, Thermo Scientific Chemicals
Vanillic acid, 98%, Thermo Scientific Chemicals
Vanillic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vanillic acid, 98%, Thermo Scientific Chemicals
CAS: 121-34-6 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA12074.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
pH4.5-7.0 (10%, demin water)
FormViscous liquid
Hydroxyl Value267- 295 mg KOH/g
View more
Vanillic acid is used as a flavoring agent in food. It acts as an intermediate in the production of vanillin from ferulic acid. Further, it is used in wine and vinegar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Vanillic acid is used as a flavoring agent in food. It acts as an intermediate in the production of vanillin from ferulic acid. Further, it is used in wine and vinegar.
Solubility
Soluble in water, alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Grieman, M. M.; Greaves, J.; Saltzman, E. S. A method for analysis of vanillic acid in polar ice cores. Clim. Past 2015, 11 (2), 227-232.
  2. Prince, P. S. M.; Dhanasekar, K.; Rajakumar, S. Vanillic acid prevents altered ion pumps, ions, inhibits Fas-receptor and caspase mediated apoptosis-signaling pathway and cardiomyocyte death in myocardial infarcted rats. Chem. Biol. Interact. 2015, 232, 68-76.