2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals
2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals
2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals
2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals
CAS: 1072-98-6 | C5H5ClN2 | 128.559 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12063.1425 g
Catalog number ALFA12063.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-chloropyridine
CAS1072-98-6
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c