o-Phenylenediamine, 98%, Thermo Scientific Chemicals
o-Phenylenediamine, 98%, Thermo Scientific Chemicals
o-Phenylenediamine, 98%, Thermo Scientific Chemicals
o-Phenylenediamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Phenylenediamine, 98%, Thermo Scientific Chemicals
CAS: 95-54-5 | C6H8N2 | 108.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
25 g
250 g
1000 g
Catalog number ALFA11946.0I
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5000 g
Request bulk or custom format
Chemical Identifiers
CAS87-62-7
IUPAC Name2,6-dimethylaniline
Molecular FormulaC8H11N
InChI KeyUFFBMTHBGFGIHF-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to white to yellow
Refractive Index1.5590-1.5620 @ 20?C
Assay (GC)≥98.5%
o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.
Solubility
Soluble in alcohol, chloroform, ether, slightly soluble in water. Soluble in hot water.
Notes
Moisture and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chen, X.; Zhang, Q.; Qian, C.; Hao, N.; Xu, L.; Yao, C. Electrochemical aptasensor for mucin 1 based on dual signal amplification of poly (o-phenylenediamine) carrier and functionalized carbon nanotubes tracing tag. Biosens. Bioelectron. 2015, 64, 485-492.
  2. Yuan, C.; Liu, X.; Jia, M.; Luo, Z.; Yao, J. Facile preparation of N-and O-doped hollow carbon spheres derived from poly (o-phenylenediamine) for supercapacitors. J. Mater. Chem. A 2015, 3 (7), 3409-3415.