Antimony(III) chloride, 99+%, Thermo Scientific Chemicals
Antimony(III) chloride, 99+%, Thermo Scientific Chemicals
Antimony(III) chloride, 99+%, Thermo Scientific Chemicals
Antimony(III) chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Antimony(III) chloride, 99+%, Thermo Scientific Chemicals
CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA11944.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS51523-79-6
IUPAC Name(2,4-difluorophenyl)hydrazine
Molecular FormulaC6H6F2N2
InChI KeyRLZUIPTYDYCNQI-UHFFFAOYSA-N
SMILESNNC1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.
Solubility
Soluble in water, hydrochloric acid, alcohol, benzene, carbon disulfide, dioxane, chloroform, ether, acetone and carbon tetrachloride. Insoluble in organic bases.
Notes
Moisture sensitive and hygroscopic in nature. It is corrosive to metals and tissue. Incompatible with strong oxidizing agents, strong acids and strong bases. Decomposes in water.
RUO – Research Use Only
General References:
  1. Mild Lewis acid catalyst.
  2. Preferred catalyst for fluoro-Pummerer reaction with DAST (Diethyl aminosulfur trifluoride, A11992) for the formation of ɑ-fluoro thioethers from sulfoxides: Tetrahedron Lett., 29, 5729 (1988); J. Org. Chem., 55, 4757 (1990); Org. Synth. Coll., 9, 446 (1998). Direct conversion of thioethers to the ɑ-fluoro analogues has also been reported: J. Org. Chem., 58, 3800 (1993).
  3. Superior to other Lewis acids for promoting the palladium(0)-catalyzed conjugate arylation of ɑß-unsaturated ketones and aldehydes with tetraphenyl borate and arylboronic acids: J. Org. Chem., 60, 883 (1995). Effective catalyst for conversion of epoxides to thiiranes with (NH4)2SCN: Indian J. Chem., 38B, 605 (1999).
  4. Lia, W.; Rong, P.; Yang, K.; Huang, P.; Sun, K.; Chen, X. Semimetal nanomaterials of antimony as highly efficient agent for photoacoustic imaging and photothermal therapy. Biomaterials 2015, 45, 18-26.
  5. Li, W.; Rong, P.; Yang, K.; Huang, P.; Sun, K.; Chen, X. Semimetal nanomaterials of antimony as highly efficient agent for photoacoustic imaging and photothermal therapy. Biomaterials 2015, 45, 18-26.
  6. Zhou, B.; Li, M.; Wu, Y.; Yang, C.; Zhang, W. H.; Li, C. Monodisperse AgSbS2 Nanocrystals: Size-Control Strategy, Large-Scale Synthesis, and Photoelectrochemistry. Chem. Eur. J. 2015, 21 (31), 11143-11151.