Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals
Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals
Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals
Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals
CAS: 68-05-3 | C8H20IN | 257.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11783.1850 g
Catalog number ALFA11783.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethylammonium iodide
CAS68-05-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.
Solubility
Soluble in water almost transparency.
Notes
Light Sensitive and Hygroscopic. Store in dark and away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only
General References:
  1. Samuel J Goodchild; Hongjian Xu; Zeineb Es-Salah-Lamoureux; Christopher A Ahern; David Fedida. Basis for allosteric open-state stabilization of voltage-gated potassium channels by intracellular cations. Journal of General Physiology, 2012, 140 (5), 495-511.
  2. Ling Xiao et. al. Unique cardiac Purkinje fiber transient outward current β-subunit composition: a potential molecular link to idiopathic ventricular fibrillation. Circulation Research, 2013, 112 (10), 1310-1322.
  3. Reagent for conversion of ɑß-enones in TFA to ß-iodo ketones in high yield: Tetrahedron, 39, 1529 (1983).