1-Pentylamine, 98%, Thermo Scientific Chemicals
1-Pentylamine, 98%, Thermo Scientific Chemicals
1-Pentylamine, 98%, Thermo Scientific Chemicals
1-Pentylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Pentylamine, 98%, Thermo Scientific Chemicals
CAS: 110-58-7 | C5H13N | 87.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA11674.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6192-52-5
IUPAC Name4-methylbenzene-1-sulfonic acid hydrate
Molecular FormulaC7H10O4S
InChI KeyKJIFKLIQANRMOU-UHFFFAOYSA-N
SMILESO.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with CHA>=98.5 % (On dry substance)
Water10.0 to 13.0 % (KF)
Appearance (Form)Adhering crystals
Infrared spectrumConforms
Titration with CHA=<0.5 % Sulfuric acid
View more
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.
Solubility
Soluble in alcohol, ether.
Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Patrik Spanel.; David Smith. Selected ion flow tube studies of the reactions of H3O+, NO+, and O2+ with eleven amine structural isomers of c5h13n1. International Journal of Mass Spectrometry. 1999, 185-187, 139-147.