3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA11664.22100 g
Catalog number ALFA11664.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Density(20/20°C) 0.865 to 0.870
Boiling point109°C to 111°C
Residue after evaporation=<0.001 %
Sulfur compounds (as S)=<0.003 %
View more
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.
Solubility
Soluble in 50 mg/mL, 95% Ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Norbert Hoffmann.; Jean-Pierre Pete. Acid catalyzed intramolecular [2+2] photocycloaddition of 3,5-dihydroxybenzoic acid derivatives. Tetrahedron Letters. 1998, 39, (28), 5027-5030.