Aluminum acetate, basic hydrate, Thermo Scientific Chemicals
Aluminum acetate, basic hydrate, Thermo Scientific Chemicals
Aluminum acetate, basic hydrate, Thermo Scientific Chemicals
Aluminum acetate, basic hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum acetate, basic hydrate, Thermo Scientific Chemicals
CAS: 142-03-0 | C4H7AlO5 | 162.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA11620.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS2591-17-5
IUPAC Name(4S)-2-[(2Z)-6-oxo-2,6-dihydro-1,3-benzothiazol-2-ylidene]-1,3-thiazolidine-4-carboxylic acid
Molecular FormulaC11H8N2O3S2
InChI KeyIWJYWBVPCGUPLO-BFUDMSGGSA-N
SMILESOC(=O)[C@H]1CSC(N1)=C1/SC2=CC(=O)C=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Appearance (Form)Powder
Water=<2.0 % (K.F.)
Specific optical rotation-32.0° to -36.0° (20°C, 589 nm) (c=1, DMF)
UV Lambda max327 to 331 nm (c=1, CH3OH)
View more
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.
Solubility
Insoluble in water
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Al-Ghamdi, A. A.; Al-Hartomy, O. A.; El Okr, M.; Nawar, A. M.; El-Gazzar, S.; El-Tantawy, F.; Yakuphanoglu, F. Semiconducting properties of Al doped ZnO thin films. Spectrochim. Acta, Part A 2014, 131, 512-517.
  2. Shin, H. U.; Stefaniak, A. B.; Stojilovic, N.; Chase, G. G. Comparative dissolution of electrospun Al2O3 nanofibres in artificial human lung fluids. Environ. Sci.: Nano 2015, 2 (3), 251-261.