Potassium benzoate, 99%, Thermo Scientific Chemicals
Potassium benzoate, 99%, Thermo Scientific Chemicals
Potassium benzoate, 99%, Thermo Scientific Chemicals
Potassium benzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium benzoate, 99%, Thermo Scientific Chemicals
CAS: 582-25-2 | C7H5KO2 | 160.213 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11610.30250 g
Catalog number ALFA11610.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS88189-03-1
IUPAC Namebismuth(3+) tritrifluoromethanesulfonate
Molecular FormulaC3BiF9O9S3
InChI KeyNYENCOMLZDQKNH-UHFFFAOYSA-K
SMILES[Bi+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (unspecified)≥98.5% on basis of Fluoride EA (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Potassium Benzoate is manufactured primarily for food and beverage use. It is a chemical preservative, which in very low concentrations inhibits the activity of the microorganisms. It is used in carbonated beverages. The shelf life of un-pasteurized cider can be greatly extended by adding potassium benzoate. It is also used as the whistle in many fireworks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium Benzoate is manufactured primarily for food and beverage use. It is a chemical preservative, which in very low concentrations inhibits the activity of the microorganisms. It is used in carbonated beverages. The shelf life of un-pasteurized cider can be greatly extended by adding potassium benzoate. It is also used as the whistle in many fireworks.
Solubility
Soluble in ethanol, slightly soluble in methanol, insoluble in ether.
Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
RUO – Research Use Only
General References:
  1. Douglas T. Gjerde; James S. Fritz. Sodium and potassium benzoate and benzoic acid as eluents for ion chromatography. Anal. Chem.1981, 53 (14), 2324-2327.
  2. M.V. Gokhale; A.T. Naik; L.K. Doraiswamy. Gas-solid reactions: an unusual observation in the disproportionation of potassium benzoate to terephthalate. Chemical Engineering Science.1975, 30 1409-1414.