2,3-Dichloroaniline, 99%, Thermo Scientific Chemicals
2,3-Dichloroaniline, 99%, Thermo Scientific Chemicals
2,3-Dichloroaniline, 99%, Thermo Scientific Chemicals
2,3-Dichloroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Dichloroaniline, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA11524.22100 g
Catalog number ALFA11524.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS26311-45-5
IUPAC Name4-pentylbenzoic acid
Molecular FormulaC12H16O2
InChI KeyCWYNKKGQJYAHQG-UHFFFAOYSA-N
SMILESCCCCCC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)84.0-91.0?C
FormPowder
Assay (Silylated GC)≥98.5%
2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.
Solubility
Soluble in ether, alcohol and acetone. Slightly soluble in benzene. Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only