1-Chlorobutane, 99+%, Thermo Scientific Chemicals
1-Chlorobutane, 99+%, Thermo Scientific Chemicals
1-Chlorobutane, 99+%, Thermo Scientific Chemicals
1-Chlorobutane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chlorobutane, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALFA11494.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS5390-04-5
IUPAC Namepent-4-yn-1-ol
Molecular FormulaC5H8O
InChI KeyCRWVOXFUXPYTRK-UHFFFAOYSA-N
SMILESOCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥94.0%
Water Content (Karl Fischer Titration)≤1.5%
Refractive Index1.4425-1.4475 @ 20°C
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.
Solubility
Miscible with alcohol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. The Grignard reagent has been recommended as superior to EtMgBr for the preparation of ethynyl magnesium halides from acetylene. For details, see: Org. Synth. Coll., 8, 606 (1993); for generation and use of the Grignard in the Mn-Cu catalyzed 1,4-addition to an enone, see: Org. Synth. Coll., 9, 328 (1998).
  2. Mututuvari, T. M.; Tran, C. D. Volumetric Study for the Binary Nitromethane with Chloroalkane Mixtures at Temperatures in the Range (298.15 to 318.15) K. J. Hazard. Mater. 2014, 264, 449-459.
  3. Atanase, L. I.; Bistac, S.; Riess, G. Effect of poly(vinyl alcohol-co-vinyl acetate) copolymer blockiness on the dynamic interfacial tension and dilational viscoelasticity of polymer-anionic surfactant complex at the water-1-chlorobutane interface. Soft matter 2015, 11 (13), 2665-2672.