Diethyl adipate, 99%, Thermo Scientific Chemicals
Diethyl adipate, 99%, Thermo Scientific Chemicals
Diethyl adipate, 99%, Thermo Scientific Chemicals
Diethyl adipate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl adipate, 99%, Thermo Scientific Chemicals
CAS: 141-28-6 | C10H18O4 | 202.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11476.22100 g
Catalog number ALFA11476.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethyl adipate
CAS141-28-6
Melting Point-20°C
Recommended StorageAmbient temperatures
Density1.008
View more
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Asok K. Dikshit,; Arun K. Nandi.Thermoreversible Gelation of Poly(vinylidene fluoride) in Diethyl Adipate: A Concerted Mechanism . Macromolecules,. 1998, 38 (25), 8886-8892.
  2. Fumio Toda,; Takaaki Suzuki,; Satoru Higa. Solvent-free Dieckmann condensation reactions of diethyl adipate and pimelate . J. Chem. Soc., Perkin Trans. 1. 1998, (21), 3521-3522.