Carbazole, 95%, Thermo Scientific Chemicals
Carbazole, 95%, Thermo Scientific Chemicals
Carbazole, 95%, Thermo Scientific Chemicals
Carbazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 95%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number ALFA11448.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS393-85-1
IUPAC Namemethyl 5-fluoro-2-nitrobenzoate
Molecular FormulaC8H6FNO4
InChI KeyLCAUVFVOLNRVKG-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C=CC(F)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.
Solubility
It is insoluble in water.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Bjorn Akermark.; Lennart Eberson.; Ernst Jonsson.; Egon Pettersson. Palladium-promoted cyclization of diphenyl ether, diphenylamine, and related compounds. J. Org. Chem.197540, (9), 1365-1367.
  2. Frederick G. Bordwell.; George E. Drucker.; Herbert E. Fried. Acidities of carbon and nitrogen acids: the aromaticity of the cyclopentadienyl anion. J. Org. Chem.198146, (3), 632-635.