Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals
Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals
Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals
Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number ALFA11316.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Heavy metals (as Pb)=<1 ppm
Free acid (CH3COOH)=<0.002 %
Density(20/4°C) 1.224 to 1.232 g/ml
View more
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates
Solubility
Soluble in water.(1 M at 20°C)
Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
RUO – Research Use Only
L Bohaty; S Haussühl. Elastic, electrostrictive, and electro-optic properties of orthorhombic sodium dihydrogen phosphate dihydrate. Zeitschrift für Kristallographie-Crystalline Materials.1984, 167, 303-306.