Barium nitrate, 99%, Thermo Scientific Chemicals
Barium nitrate, 99%, Thermo Scientific Chemicals
Barium nitrate, 99%, Thermo Scientific Chemicals
Barium nitrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Barium nitrate, 99%, Thermo Scientific Chemicals
CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11305.36500 g
Catalog number ALFA11305.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormRod
Diameter2.7 to 3.3 mm
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Moisture sensitive. Incompatible with metal,alloy powders, combustible materials, reducing agents, acids, and acid anhydrides.
RUO – Research Use Only
General References:
  1. Lu, F.; Zhao, B.; Li, R.; Ruan, W. Crystal growth of barium nitrate on thiol-terminated self-assembled monolayers and a Raman spectroscopic investigation of the crystal facets. J. Cryst. Growth. 2015, 426, 33-37.
  2. Bardwell, C. J.; Bickley, R. I.; Poulston, S.; Twigg, M. V. Thermal Decomposition of bulk and supported Barium Nitrate. Thermochim acta. 2015, 613, 94-99.