(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
CAS: 13822-56-5 | C6H17NO3Si | 179.291 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA11284.36
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS556-50-3
IUPAC Namehydrogen 2-(2-aminoacetamido)acetic acid chloride
Molecular FormulaC4H9ClN2O3
InChI KeyYHBAZQDEMYQPJL-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Heavy metals (as Pb)=<5 ppm
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C, 4 h)
pH5.5 to 6.5 (5 % in water)
View more
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).
Solubility
Miscible with water.
Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
RUO – Research Use Only
General References:
  1. Schalken, J. R. G.; Creatore, M.; Verhoeven, P.; Stevens, A. mu Plasma Printing Deposition of Amine-Containing Organo-Silane Polymers by Means of 3-Aminopropyl Trimethoxysilane. Nanosci. Nanotechnol. Lett. 2015, 7 (1), 62-66.
  2. Jones, S.; Kolpin, A.; Tsang, S. C. E. Modification of Pd for formic acid decomposition by support grafted functional groups. Catal. Struct. React. 2015, 1 (1), 19-24.