4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals
4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals
4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals
4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA11200.22100 g
Catalog number ALFA11200.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS12033-81-7
IUPAC Namedihydrogen dinitroplatinumbis(ylium) sulfate
Molecular FormulaH2N2O8PtS
InChI KeySPPCQWREVYSZTJ-UHFFFAOYSA-N
SMILES[H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)Pt 4-6%